Learn More
Biomatik Corporation Human Interleukin 8 Receptor Beta (IL8Rb) ELISA Kit, 96 tests
Catalog No. 501503042
Encompass_Preferred
The Human Interleukin 8 Receptor Beta (IL8Rb) ELISA Kit allows for quantitative determination interleukin 8 Receptor Beta in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25025. Cross Activity: No significant cross-reactivity or interference between Interleukin 8 Receptor Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3042 Supplier Biomatik Corporation Supplier No. EKU05387
The Human Interleukin 8 Receptor Beta (IL8Rb) ELISA Kit allows for quantitative determination interleukin 8 Receptor Beta in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P25025. Cross Activity: No significant cross-reactivity or interference between Interleukin 8 Receptor Beta and analogues was observed. Restriction: For Research Use Only.