Learn More
Biomatik Corporation Human Actin, Cytoplasmic 2 (Actg1) Elisa Kit
Supplier:  Biomatik Corporation EKC32468
Encompass_Preferred
Human Actin, Cytoplasmic 2 (Actg1) Elisa Kit for detecting Actin, Gamma 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P63261; ACTG1. Target Synonyms: ACT; ACTB; ACTG; ACTG_HUMAN; actg1; Actin; Actin; cytoplasmic 2; Actin; gamma 1; Actin; gamma 1 propeptide; Actin; gamma; BRWS2; cytoplasmic 2; Cytoskeletal gamma actin; Deafness; autosomal dominant 20; Deafness; autosomal dominant 26; DFNA20; DFNA26; epididymis luminal protein 176; Gamma-actin; HEL-176; N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7303