Learn More
Biomatik Corporation Human Adam Metallopeptidase With Thrombospondin Type 1 Motif4, Adamts4 Elisa Kit
Supplier:  Biomatik Corporation EKC32486
Encompass_Preferred
Human Adam Metallopeptidase With Thrombospondin Type 1 Motif4, Adamts4 Elisa Kit for detecting Adam Metallopeptidase With Thrombospondin Type 1 Motif4, Adamts4 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-800ng/ml. Sensitivity: 3.125ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O75173; ADAMTS4. Target Synonyms: A disintegrin and metalloproteinase with thrombospondin motifs 4; A disintegrin like and metalloprotease (reprolysin type) with thrombospondin type 1 motif 4; A disintegrin like and metalloprotease with thrombospondin type 1 motif 4; ADAM metallopeptidase with thrombospondin type 1 motif 4; ADAM TS 4; ADAM TS4; ADAM-TS 4; ADAM-TS4; ADAMTS 2; ADAMTS 4; ADAMTS-4; ADAMTS2; ADAMTS4; ADAMTS4 protein; ADMP 1; ADMP-1; ADMP1; Aggrecanase 1; Aggrecanase-1; Aggrecanase1; ATS4_HUMAN; KIAA0688. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7305