Learn More
Biomatik Corporation Rat Interleukin 9 (IL9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05392
Encompass_Preferred
Rat Interleukin 9 (IL9) ELISA Kit for detecting IL9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A8I9. Gene ID: 116558. Target Synonyms: HP40; P40; TCGFIII; TCGF3; T Cell Growth Factor III; P40 T-Cell And Mast Cell Growth Factor; P40 Cytokine; Homolog Of Mouse T Cell And Mast Cell Growth Factor 40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3044