Learn More
Biomatik Corporation Human amyloid beta peptide 1-40,Aβ1-40 ELISA Kit
Supplier:  Biomatik Corporation EKC32565
Encompass_Preferred
Sample Type: serum, tissue homogenates, cerebrospinal fluid (CSF). Species Reactivity: Homo sapiens (Human). Detection Range: 125 pg/mL-8000 pg/mL. Sensitivity: 31.25 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: amyloid beta peptide 1-40,Aβ1-40,amyloid β peptide 1-40. Restriction: For Research Use Only.
Catalog No. 50-228-7310