Learn More
Biomatik Corporation Human Islet Amyloid Polypeptide (IAPP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05403
Encompass_Preferred
The Human Islet Amyloid Polypeptide (IAPP) ELISA Kit allows for quantitative determination of Islet Amyloid Polypeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 6.17-500 pg/mL. Sensitivity: 2.55 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P10997. Cross Activity: No significant cross-reactivity or interference between Islet Amyloid Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3046