Learn More
Biomatik Corporation Human Anterior Gradient Protein 3 Homolog (Agr3) Elisa Kit
Supplier:  Biomatik Corporation EKC32610
Encompass_Preferred
Human Anterior Gradient Protein 3 Homolog (Agr3) Elisa Kit for detecting Anterior Gradient Homolog 3 (Xenopus Laevis) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8TD06; AGR3. Target Synonyms: AG-3; AG3; Agr3; AGR3_HUMAN; Anterior gradient 3; xenopus; homolog of; Anterior gradient homolog 3; Anterior gradient homolog 3 (Xenopus laevis); anterior gradient protein 3; Anterior gradient protein 3 homolog; BCMP11; Breast cancer membrane protein 11; Gm888; hAG 3; hAG-3; HAG3; OTTHUMP00000158512; PDIA18; Protein disulfide isomerase family A member 18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7320