Learn More
Biomatik Corporation Human Isocitrate Dehydrogenase 1, Soluble (IDH1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05406
Encompass_Preferred
The Human Isocitrate Dehydrogenase 1, Soluble (IDH1) ELISA Kit allows for quantitative determination of Isocitrate Dehydrogenase 1, Soluble in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.33 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75874. Cross Activity: No significant cross-reactivity or interference between Isocitrate Dehydrogenase 1, Soluble and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3048