Learn More
Biomatik Corporation Human Apelin Receptor (Aplnr) Elisa Kit
Catalog No. 502287325
Encompass_Preferred
Human Apelin Receptor (Aplnr) Elisa Kit for detecting Apelin Receptor in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35414; APLNR. Target Synonyms: APLNR; AGTRL1; APJ; Apelin receptor; Angiotensin receptor-like 1; G-protein coupled receptor APJ; G-protein coupled receptor HG11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-228-7325 Supplier Biomatik Corporation Supplier No. EKC32753
Add to Cart
Add to Cart
Human Apelin Receptor (Aplnr) Elisa Kit for detecting Apelin Receptor in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35414; APLNR. Target Synonyms: APLNR; AGTRL1; APJ; Apelin receptor; Angiotensin receptor-like 1; G-protein coupled receptor APJ; G-protein coupled receptor HG11. Restrictions: For Research Use Only. Not for use in diagnostic procedures.