Learn More
Biomatik Corporation Human Apolipoprotein C2, Apo-C2 Elisa Kit
Supplier:  Biomatik Corporation EKC32758
Encompass_Preferred
Human Apolipoprotein C2, Apo-C2 Elisa Kit for detecting Apolipoprotein C-Ii in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02655; APOC2. Target Synonyms: APC 2; APC2; Apo CII; Apo-CII; APOC 2; ApoC II; ApoC-II; APOC2; APOC2 protein; APOC2_HUMAN; ApoCII; Apolipoprotein C II; Apolipoprotein C II precursor; Apolipoprotein C2; ApolipoproteinCII; MGC75082; ProapoC-II; Proapolipoprotein C-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7327