Learn More
Biomatik Corporation Human Azurocidin/Heparin Binding Protein(AZU/HBP) ELISA Kit
Supplier:  Biomatik Corporation EKC32818
Encompass_Preferred
Sample Type: serum, urine, tissue homogenates, cell culture supernates. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P20160. Target Synonyms: AZAMP, AZU, CAP37, HBP, HUMAZUR, NAZC, cationic antimicrobial protein 37|heparin-binding protein|neutrophil azurocidin. Restriction: For Research Use Only.
Catalog No. 50-228-7335