Learn More
Biomatik Corporation Human B-Cell Leukemia/Lymphoma 6, Bcl6 Elisa Kit
Supplier:  Biomatik Corporation EKC32837
Encompass_Preferred
Human B-Cell Leukemia/Lymphoma 6, Bcl6 Elisa Kit for detecting B-Cell Cll/Lymphoma 6 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 18.75-1200pg/ml. Sensitivity: 4.68pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41182; BCL6. Target Synonyms: B cell CLL/lymphoma 6; B cell lymphoma 6 protein; B-cell lymphoma 5 protein; B-cell lymphoma 6 protein; BCL 5; Bcl 6; BCL-5; BCL-6; BCL5; BCL6; BCL6_HUMAN; BCL6A; cys his2 zinc finger transcription factor; cys-his2 zinc finger transcription factor; LAZ 3; LAZ 3 protein; LAZ3; Lymphoma Associated Zinc Finger Gene On Chromosome 3 (LAZ3); Lymphoma associated zinc finger gene on chromosome 3; Protein LAZ-3; ZBTB 27; ZBTB27; Zinc finger and BTB domain containing protein 27. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7337