Learn More
Biomatik Corporation Human Junctional Adhesion Molecule 3 (JAM3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05423
Encompass_Preferred
The Human Junctional Adhesion Molecule 3 (JAM3) ELISA Kit allows for quantitative determination of Junctional Adhesion Molecule 3 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.62-1000 pg/mL. Sensitivity: 6.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BX67. Cross Activity: No significant cross-reactivity or interference between Junctional Adhesion Molecule 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3055