Learn More
Biomatik Corporation Mouse Kallikrein 1 (KLK1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05429
Encompass_Preferred
Mouse Kallikrein 1 (KLK1) ELISA Kit for detecting KLK1 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 49.4-4,000pg/ml. Sensitivity: 19.6pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P15947. Gene ID: 16612. Target Synonyms: KLKR; Klk6; HK1; Kallikrein 1,Renal/Pancreas/Salivary; Kidney/pancreas/salivary gland kallikrein; Tissue kallikrein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3056