Learn More
Biomatik Corporation Human Bleomycin Hydrolase (Blmh) Elisa Kit
Supplier:  Biomatik Corporation EKC32882
Encompass_Preferred
Human Bleomycin Hydrolase (Blmh) Elisa Kit for detecting Bleomycin Hydrolase in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 28-1800pg/ml. Sensitivity: 7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13867; BLMH. Target Synonyms: BH; Bleomycin hydrolase; BLM hydrolase; BLMH; BLMH_HUMAN; BMH; EC 3.4.22.40. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7342