Learn More
Biomatik Corporation Human Kallikrein 3 (KLK3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05438
Encompass_Preferred
The Human Kallikrein 3 (KLK3) ELISA Kit allows for quantitative determination of Kallikrein 3 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.20-12.5 ng/mL. Sensitivity: 0.08 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07288. Cross Activity: No significant cross-reactivity or interference between Kallikrein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3058