Learn More
Biomatik Corporation Human Bone Morphogenetic Protein Receptor Ii, Bmpr-Ii Elisa Kit
Supplier:  Biomatik Corporation EKC32900
Encompass_Preferred
Human Bone Morphogenetic Protein Receptor Ii, Bmpr-Ii Elisa Kit for detecting Bone Morphogenetic Protein Receptor, Type Ii (Serine/Threonine Kinase) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13873; BMPR2. Target Synonyms: BMP type II receptor; BMP type-2 receptor; BMPR 2; BMPR 3; BMPR II; BMPR-2; BMPR-II; Bmpr2; BMPR2_HUMAN; BMPR3; BMPRII; BMR 2; BMR2; Bone morphogenetic protein receptor type 2; Bone morphogenetic protein receptor type II; Bone morphogenetic protein receptor type-2; Bone morphogenic protein receptor type II serine threonine kinase; BRK 3; BRK3; PPH 1; PPH1; Serine threonine kinase type II activin receptor like kinase; T ALK; TALK; Type II activin receptor like kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7346