Learn More
Biomatik Corporation Human Catestatin Elisa Kit
Supplier:  Biomatik Corporation EKC33038
Encompass_Preferred
Human Catestatin Elisa Kit for detecting Catestatin in serum, plasma, cell culture supernates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7356