Learn More
Biomatik Corporation Mouse Kallikrein 7 (KLK7) ELISA Kit, 96 tests
Catalog No. 501503063
Encompass_Preferred
Mouse Kallikrein 7 (KLK7) ELISA Kit for detecting KLK7 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91VE3. Gene ID: 23993. Target Synonyms: PRSS6; SCCE; hK7; Kallikrein-Related Peptidase 7; Serine protease 6; Stratum corneum chymotryptic enzyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3063 Supplier Biomatik Corporation Supplier No. EKU05450
Mouse Kallikrein 7 (KLK7) ELISA Kit for detecting KLK7 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91VE3. Gene ID: 23993. Target Synonyms: PRSS6; SCCE; hK7; Kallikrein-Related Peptidase 7; Serine protease 6; Stratum corneum chymotryptic enzyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.