Learn More
Biomatik Corporation Human Karyopherin Alpha 2 (KPNa2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05456
Encompass_Preferred
The Human Karyopherin Alpha 2 (KPNa2) ELISA Kit allows for quantitative determination of Karyopherin Alpha 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-8000 pg/mL. Sensitivity: 52 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P52292. Cross Activity: No significant cross-reactivity or interference between Karyopherin Alpha 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3064