Learn More
Biomatik Corporation Human Collagen Alpha-3 (Iv) Chain (Col4A3) Elisa Kit
Supplier:  Biomatik Corporation EKC33214
Encompass_Preferred
Human Collagen Alpha-3 (Iv) Chain (Col4A3) Elisa Kit for detecting Collagen, Type Iv, Alpha 3 (Goodpasture Antigen) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q01955; COL4A3. Target Synonyms: Alpha 3 type IV collagen; Alpha3 type IV collagen; CO4A3_HUMAN; COL4A 3; Col4a3; Collagen alpha 3 (IV) chain; Collagen IV alpha 3 polypeptide; Collagen type IV alpha 3 (Goodpasture antigen); Collagen type IV alpha 3; Collagen type IV alpha 3 chain; Goodpasture antigen; OTTHUMP00000195044; Tumstatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7369