Learn More
Biomatik Corporation Human Keratin 1 (KRT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05459
Encompass_Preferred
The Human Keratin 1 (KRT1) ELISA Kit allows for quantitative determination of Keratin 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.060 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P04264. Cross Activity: No significant cross-reactivity or interference between Keratin 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3066