Learn More
Biomatik Corporation Human Collagen Type Iv (Col Iv) Elisa Kit
Supplier:  Biomatik Corporation EKC33226
Encompass_Preferred
Human Collagen Type Iv (Col Iv) Elisa Kit for detecting Collagen Type IV(Col IV) in serum, plasma, cell culture supernates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: Collagen Type 4 (COL 4); Collagen Type IV (COL IV). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7372