Learn More
Biomatik Corporation Human Keratin 10 (KRT10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05461
Encompass_Preferred
The Human Keratin 10 (KRT10) ELISA Kit allows for quantitative determination of Keratin 10 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 11.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P13645. Cross Activity: No significant cross-reactivity or interference between Keratin 10 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3067