Learn More
Biomatik Corporation Human Interleukin 1Β, Il-1Β Elisa Kit
Supplier:  Biomatik Corporation EKC34230
Encompass_Preferred
Human Interleukin 1Β, Il-1Β Elisa Kit for detecting Interleukin 1, Beta in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01584; IL1B. Target Synonyms: Catabolin; H1; IFN beta inducing factor; IL 1; IL 1 beta; IL-1 beta; IL1; IL1 BETA; IL1B; IL1B_HUMAN; IL1F2; Interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1; beta; Interleukin-1 beta; OAF; Osteoclast activating factor; OTTHUMP00000162031; Preinterleukin 1 beta; Preinterleukin beta; Pro interleukin 1 beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7453