Learn More
Biomatik Corporation Human Keratin 18 (KRT18) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05466
Encompass_Preferred
The Human Keratin 18 (KRT18) ELISA Kit allows for quantitative determination of Keratin 18 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.060 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05783. Cross Activity: No significant cross-reactivity or interference between Keratin 18 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3069