Learn More
Biomatik Corporation Human Klotho Elisa Kit
Supplier:  Biomatik Corporation EKC34354
Encompass_Preferred
Human Klotho Elisa Kit for detecting Klotho in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9UEF7; KL. Target Synonyms: Kl; KLOT_HUMAN; Klotho peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7463