Learn More
Biomatik Corporation Human Neurofilament Protein H (Nf-H) Elisa Kit
Supplier:  Biomatik Corporation EKC34780
Encompass_Preferred
Human Neurofilament Protein H (Nf-H) Elisa Kit for detecting Neurofilament, Heavy Polypeptide in serum, plasma, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P12036; NEFH. Target Synonyms: 200 kDa neurofilament protein; CMT2CC; Nefh; Neurofilament heavy polypeptide 200kDa; Neurofilament heavy polypeptide; Neurofilament triplet H protein; NF H; NF-H; NFH; NFH_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7490