Learn More
Biomatik Corporation Human β-galactosidase,βGAL ELISA Kit
Supplier:  Biomatik Corporation EKC36117
Encompass_Preferred
Sample Type: serum, plasma, urine, tissue homogenates. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8 μlU/mL-500 μlU/mL. Sensitivity: 1.95 μlU/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P16278. Target Synonyms: EBP, ELNR1, elastin receptor 1, 67kDa,β-galactosidase,βGAL,beta-galactosidase,beta GAL. Restriction: For Research Use Only.
Catalog No. 50-228-7556