Learn More
Biomatik Corporation Mouse Homocysteic acid,Hcy ELISA Kit
Supplier:  Biomatik Corporation EKC37067
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6 pmol/mL-1000 pmol/mL. Sensitivity: 3.9 pmol/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7598