Learn More
Biomatik Corporation Mouse Lactoferrin, Ltf/Lf Elisa Kit
Supplier:  Biomatik Corporation EKC37252
Encompass_Preferred
Mouse Lactoferrin, Ltf/Lf Elisa Kit for detecting Lactotransferrin in serum, plasma, breast milk. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ug/ml. Sensitivity: 3.12ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P08071; Ltf. Target Synonyms: GIG12; HLF2; LF; growth-inhibiting protein 12; lactoferrin; neutrophil lactoferrin; talalactoferrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7608