Learn More
Biomatik Corporation Human Kidney Injury Molecule 1 (Kim1) ELISA Kit, 96 tests
Catalog No. 501503080
Encompass_Preferred
The Human Kidney Injury Molecule 1 (Kim1) ELISA Kit allows for quantitative determination of Kidney Injury Molecule 1 in Tissue homogenates, Urine, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5,000 pg/mL. Sensitivity: 28 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96D42. Cross Activity: No significant cross-reactivity or interference between Kidney Injury Molecule 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3080 Supplier Biomatik Corporation Supplier No. EKU05489
The Human Kidney Injury Molecule 1 (Kim1) ELISA Kit allows for quantitative determination of Kidney Injury Molecule 1 in Tissue homogenates, Urine, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5,000 pg/mL. Sensitivity: 28 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q96D42. Cross Activity: No significant cross-reactivity or interference between Kidney Injury Molecule 1 and analogues was observed. Restriction: For Research Use Only.