Learn More
Biomatik Corporation Mouse Pigment Epithelium-Derived Factor, Pedf Elisa Kit
Supplier:  Biomatik Corporation EKC37578
Encompass_Preferred
Mouse Pigment Epithelium-Derived Factor, Pedf Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade F (Alpha-2 Antiplasmin, Pigment Epithelium Derived Factor), Member 1 in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P97298; Serpinf1. Target Synonyms: Serpinf1; Pedf; Sdf3Pigment epithelium-derived factor; PEDF; Caspin; Serpin F1; Stromal cell-derived factor 3; SDF-3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7617