Learn More
Biomatik Corporation Rat Kidney Injury Molecule 1 (Kim1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05491
Encompass_Preferred
Rat Kidney Injury Molecule 1 (Kim1) ELISA Kit for detecting Kim1 in urine, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 24.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O54947. Gene ID: 286934. Target Synonyms: HAVCR1; TIM1; TIMD1; HAVCR; Hepatitis A Virus Cellular Receptor 1; T Cell Immunoglobulin And Mucin Domain-Containing Protein 1; T-cell immunoglobulin mucin receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3082