Learn More
Biomatik Corporation Rat Complement 3,C3 ELISA Kit
Supplier:  Biomatik Corporation EKC38878
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625 μg/mL-40 μg/mL. Sensitivity: 0.156 μg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01026. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7653