Learn More
Biomatik Corporation Rat Retinol binding protein 4,RBP-4 ELISA Kit
Supplier:  Biomatik Corporation EKC39649
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 39 ng/ml-2500 ng/ml.. Sensitivity: 39 ng/ml.. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04916. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7679