Learn More
Biomatik Corporation Human Activated Leukocyte Cell Adhesion Molecule, Alcam Elisa Kit
Supplier:  Biomatik Corporation EKC40024
Encompass_Preferred
Human Activated Leukocyte Cell Adhesion Molecule, Alcam Elisa Kit for detecting Activated Leukocyte Cell Adhesion Molecule in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.039-2.5ng/ml. Sensitivity: 0.0097ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q13740; ALCAM. Target Synonyms: Activated leukocyte cell adhesion molecule; ALCAM; ALCAM protein; CD 166; CD166; CD166 antigen; CD166_HUMAN; FLJ3851; FLJ38514; MEMD; MGC71733. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7689