Learn More
Biomatik Corporation Human Neutrophil Activating Protein-2, Nap-2 Elisa Kit
Supplier:  Biomatik Corporation EKC40048
Encompass_Preferred
Human Neutrophil Activating Protein-2, Nap-2 Elisa Kit for detecting Neutrophil-Activating Peptide 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02775; PPBP. Target Synonyms: B TG1; Beta TG; Beta thromboglobulin; Beta-TG; C-X-C motif chemokine 7; Chemokine (C X C motif) ligand 7; Connective tissue activating peptide III; CTAP 3; CTAP III; CTAP-III; CTAP-III (1-81); CTAP3; CTAPIII; CXC chemokine ligand 7; CXCL 7; CXCL7; CXCL7_HUMAN; LA PF 4; LA-PF4; LDGF; Leukocyte derived growth factor; Leukocyte-derived growth factor; Low-affinity platelet factor IV; Macrophage-derived growth factor; MDGF; NAP 2; NAP-2; NAP-2 (1-63); NAP-2 (1-66); NAP-2 (73); NAP-2 (74). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7696