Learn More
Biomatik Corporation Human Klotho (KL) ELISA Kit, 96 tests
Catalog No. 501503087
Encompass_Preferred
The Human Klotho (KL) ELISA Kit allows for quantitative determination of Klotho in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UEF7. Cross Activity: No significant cross-reactivity or interference between Klotho and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3087 Supplier Biomatik Corporation Supplier No. EKU05510
The Human Klotho (KL) ELISA Kit allows for quantitative determination of Klotho in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9UEF7. Cross Activity: No significant cross-reactivity or interference between Klotho and analogues was observed. Restriction: For Research Use Only.