Learn More
Biomatik Corporation Rat Laminin,LN ELISA kit
Supplier:  Biomatik Corporation EKC40090
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 62.5 pg/mL-4000 pg/mL. Sensitivity: 15.6 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-228-7704