Learn More
Biomatik Corporation Mouse P-Selectin ELISA kit
Supplier:  Biomatik Corporation EKC40111
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q01102. Target Synonyms: RP1-86F14.2, CD62, CD62P, FLJ45155, GMP140, GRMP, LECAM3, PADGEM, PSEL, OTTHUMP00000032609|granulocyte membrane protein|leukocyte-endothelial cell adhesion molecule 3|platelet alpha-granule membrane. Restriction: For Research Use Only.
Catalog No. 50-228-7709