Learn More
Biomatik Corporation Human Kruppel Like Factor 4, Gut (KLF4) ELISA Kit, 96 tests
Catalog No. 501503090
Encompass_Preferred
The Human Kruppel Like Factor 4, Gut (KLF4) ELISA Kit allows for quantitative determination of Kruppel Like Factor 4, Gut in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43474. Cross Activity: No significant cross-reactivity or interference between Kruppel Like Factor 4, Gut and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3090 Supplier Biomatik Corporation Supplier No. EKU05519
The Human Kruppel Like Factor 4, Gut (KLF4) ELISA Kit allows for quantitative determination of Kruppel Like Factor 4, Gut in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.056 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O43474. Cross Activity: No significant cross-reactivity or interference between Kruppel Like Factor 4, Gut and analogues was observed. Restriction: For Research Use Only.