Learn More
Biomatik Corporation Mouse myelin basic protein,MBP ELISA Kit
Supplier:  Biomatik Corporation EKC40423
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 18.75 pg/mL-1200 pg/mL. Sensitivity: 4.7 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P04370. Target Synonyms: MGC99675, Golli-mbp|OTTHUMP00000174383. Restriction: For Research Use Only.
Catalog No. 50-228-7733