Learn More
Biomatik Corporation Human Lactate Dehydrogenase A (LDHA) ELISA Kit, 96 tests
Catalog No. 501503093
Encompass_Preferred
The Human Lactate Dehydrogenase A (LDHA) ELISA Kit allows for quantitative determination of Lactate Dehydrogenase A in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00338. Cross Activity: No significant cross-reactivity or interference between Lactate Dehydrogenase A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3093 Supplier Biomatik Corporation Supplier No. EKU05532
The Human Lactate Dehydrogenase A (LDHA) ELISA Kit allows for quantitative determination of Lactate Dehydrogenase A in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.067 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00338. Cross Activity: No significant cross-reactivity or interference between Lactate Dehydrogenase A and analogues was observed. Restriction: For Research Use Only.