Learn More
Biomatik Corporation Human Procalcitonin, Pct Elisa Kit
Supplier:  Biomatik Corporation EKC40634
Encompass_Preferred
Human Procalcitonin, Pct Elisa Kit for detecting Procalcitonin, Pct in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7765