Learn More
Biomatik Corporation Monkey Interferon γ ,IFN-γ ELISA Kit
Supplier:  Biomatik Corporation EKC40734
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Monkey. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.8 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P63310. Target Synonyms: IFG, IFI,IFN-gamma,interferon gamma. Restriction: For Research Use Only.
Catalog No. 50-228-7776