Learn More
Biomatik Corporation Cattle Lactoferrin (LTF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05537
Encompass_Preferred
The Cattle Lactoferrin (LTF) ELISA Kit allows for quantitative determination of Lactoferrin in Serum, Plasma, Tissue homogenates, Cell lysates, Saliva, milk, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.64 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P24627. Cross Activity: No significant cross-reactivity or interference between Lactoferrin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3095