Learn More
Biomatik Corporation Human Nuclear Factor-Kappa B, Nf-Κb Elisa Kit
Supplier:  Biomatik Corporation EKC40846
Encompass_Preferred
Human Nuclear Factor-Kappa B, Nf-Κb Elisa Kit for detecting Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P19838; NFKB1. Target Synonyms: DKFZp686C01211; DNA binding factor KBF1; DNA binding factor KBF1 EBP1; DNA-binding factor KBF1; EBP 1; EBP-1; EBP1; KBF1; MGC54151; nf b; NF kappa B; NF kappaB; NF kappabeta; NF kB1; NF-kappaB; NFkappaB; NFKB 1; NFKB p105; NFKB p50; NFKB-p105; Nfkb1; NFKB1_HUMAN; Nuclear factor kappa B DNA binding subunit; Nuclear factor kappa-B; Nuclear factor kappa-B; subunit 1; Nuclear factor NF kappa B p105 subunit; Nuclear factor NF kappa B p50 subunit; Nuclear factor NF-kappa-B p50 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7791