Learn More
Biomatik Corporation Human Lactoferrin (LTF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05538
Encompass_Preferred
The Human Lactoferrin (LTF) ELISA Kit allows for quantitative determination of Lactoferrin in Serum, Plasma, Tissue homogenates, Cell lysates, Breast milk, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.135 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02788. Cross Activity: No significant cross-reactivity or interference between Lactoferrin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3096