Learn More
Biomatik Corporation Mouse Interleukin 21 (Il-21) Elisa Kit
Supplier:  Biomatik Corporation EKC40952
Encompass_Preferred
Mouse Interleukin 21 (Il-21) Elisa Kit for detecting Interleukin 21 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9ES17; Il21. Target Synonyms: Il21Interleukin-21; IL-21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-7803